Learn More
Medchemexpress LLC Rubrofusarin gentiobioside | 24577-90-0 | 99.0% | 596.53 g/mol | C27H32O15 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN417710MG
Encompass_Preferred
Rubrofusarin gentiobioside is a naturally occurring phenolic glycoside isolated from the seeds of Cassia tora L. It is supplied for research use and has been reported to exhibit radical scavenging (antioxidant) activity. The product is provided with analytical documentation to support purity and safe handling.
  • Isolated from seeds of Cassia tora L.
  • Exhibits radical scavenging activity in antioxidant assays.
  • High purity (99.04%) suitable for research applications.
  • Molecular formula C27H32O15 and molecular weight 596.53 g/mol.
  • Supplied as a solid in small pack sizes, including 10 MG.
  • Provided with COA and SDS documents for quality and safety.
Catalog No. 50-002-89952