Learn More
Medchemexpress LLC Rubusoside | 64849-39-4 | 642.73 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN066810MM/1ML
Encompass_Preferred
Rubusoside is a diterpene glycoside that functions as a sweetener and solubilizer. It is an analytical standard intended for research applications. This compound exhibits a range of biological activities including anti-angiogenic, anti-cancer, anti-obesity, anti-allergic, and anti-asthmatic effects.
  • Attenuates airway hyperresponsiveness
  • Reduces inflammatory cells in bronchoalveolar lavage fluid (BALF)
  • Alleviates OVA-induced airway inflammation
  • Prevents palmitic acid-induced lipotoxicity in pancreatic INS-1 cells
  • Decreases transport of human glucose transporters GLUT-1 and fructose GLUT-5
  • Inhibits NF-κB and α-amylase
Catalog No. 50-003-88868