Learn More
Medchemexpress LLC Rubusoside | 64849-39-4 | 642.73 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN066850MG
Encompass_Preferred
Rubusoside is a diterpene glycoside that functions as a sweetener and solubilizer. It exhibits various properties including anti-angiogenic, anti-cancer, anti-obesity, anti-allergic, and anti-asthmatic effects. It can attenuate airway hyperresponsiveness and reduce inflammatory cells in bronchoalveolar lavage fluid (BALF).
  • Mitigates OVA-induced airway inflammation.
  • Prevents palmitic acid-induced lipotoxicity in pancreatic INS-1 cells.
  • Decreases the transport of human glucose transporters GLUT-1 and fructose GLUT-5.
  • Inhibits NF-κB and α-amylase.
  • Intended for research use only.
Catalog No. 50-003-88869