Learn More
Apexbio Technology LLC Rucaparib (AG-014699,PF-01367338), 459868-92-9, MFCD17010269, 5 mg
SDP
Supplier:  Apexbio Technology LLC A41565
Encompass_Preferred
MF: C19H18FN3OH3PO4, MW: 421.36, Synonyms: Rucaparib phosphate, AG-14699, AG 014699. Solubility: 21.1 mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Rucaparib, also named as AG-014699 or PF-01367338, is a poly (ADP ribose) polymerase (PARP) inhibitor.
Catalog No. 50-101-2981