Learn More
TARGETMOL CHEMICALS INC RUFIGALLOL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T261685MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Rufigallol (AI3-00865) is an electron-deficient disk-shaped molecule with six hydroxyl groups and is a good unit for inducing the formation of DLC from non-planar AIE molecules an anthraquinone derivative.Rufigallol induces the acquisition of DLC properties while the introduction of TPE groups gives the molecular structure good fluorescence in the aggregated state.The remarkable synergistic in vitro antimalarial effect in vivo with vitamin C and ketones was synergistic and showed antimalarial activity. purity: 99%
Catalog No. 50-258-0717