Learn More
Apexbio Technology LLC Rufinamide 106308-44-5 10mg
SDP
Supplier:  Apexbio Technology LLC B228010
Encompass_Preferred
Rufinamide is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to modulate neuronal excitability by stabilizing sodium channel inactivation thereby reducing repetitive neuronal firing and seizure propagation Rufinamide exerts its biological activity primarily through inhibition of voltage-gated sodium channels Based on these pharmacological properties Rufinamide holds research potential in neuroscience studies particularly as an adjunctive therapeutic candidate for severe epilepsy subtypes such as Lennox-Gastaut syndrome and in investigations of cellular electrophysiological mechanisms underlying epileptic disorders
Catalog No. 50-224-0776