Learn More
ATCC Ruminococcus gnavus; Strain VPI C7-9
Supplier:  ATCC 29149
Encompass
Ruminococcus gnavus strain VPI C7-9 is a whole-genome sequenced type strain that was isolated from human feces. This anaerobic bacterium has been sequenced as a reference genome for the NIH Human Microbiome Project.
Catalog No. 50-238-3752
 Restricted Item