Learn More
Selleck Chemical LLC Rupatadine E7255-50MG
SDP
Supplier:  Selleck Chemical LLC E725550MG
Encompass_Preferred
Rupatadine (UR-12592) is a potent orally active and long-lasting dual PAF/H1 antagonist with Kis of 0 55 M and 0 1 M respectively Rupatadine can be used for the research of allergic rhinitis and urticaria
Catalog No. 50-000-42193