Learn More
Strem, An Ascensus Company CAS# [1028206-60-1]. 100mg. Chloro(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)[2-(2-aminoethylphenyl)]palladium(II), methyl-t-butylether adduct, min. 98% [RuPhos Palladacycle Gen. 1]. MFCD12545955
SDP
Supplier:  Strem, An Ascensus Company 460266100MG
Encompass
CAS# [1028206-60-1]. 100mg. Chloro(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)[2-(2-aminoethylphenyl)]palladium(II), methyl-t-butylether adduct, min. 98% [RuPhos Palladacycle Gen. 1]. MFCD12545955. Molecular Weight: 728.68. Molecular Formula: C38H53ClNO2PPd. Color/form: off-white to beige pwdr. Strem# 46-0266. www.strem.com/catalog/v3/46-0266/
Catalog No. 50-197-9388