Learn More
Medchemexpress LLC Ruscogenin | 874485-32-2 | 99.8% | 430.62 g/mol | C27H42O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN513610MG
Encompass_Preferred
25(R,S)-Ruscogenin is a steroidal sapogenin research compound used to modulate inflammatory and cancer-related signaling pathways. It inhibits PI3K/Akt/mTOR and TLR4 signaling and has been reported to suppress hepatocellular carcinoma metastasis and reduce LPS-induced pulmonary endothelial apoptosis.
  • Functions as a PI3K/Akt/mTOR and TLR4 signaling inhibitor.
  • Reported to reduce expression of MMP-2, MMP-9, uPA, VEGF, and HIF-1α.
  • High purity (approximately 99.8%).
  • Solid powder, white to off-white in color.
  • Available in small research pack sizes including 10 mg.
  • Identifiers: CAS 874485-32-2; formula C27H42O4; mol weight 430.62 g/mol.
  • Store sealed at 4°C, protect from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-89931