Learn More
Electron Microscopy Sciences Russian Tissue Forceps 6"
SDP
Supplier:  Electron Microscopy Sciences 7826006
Encompass_Preferred
Russian tissue forceps. Stainless steel 6" (155mm) long, with special cupped pattern for superior grip, cup area is 6.5mm wide, 9mm long.
Catalog No. 50-284-10