Learn More
Enterprise Technology Solutions Rust-Oleum Spray Paint,CorrosionProtection,15 oz,YW
SDP
Supplier:  Enterprise Technology Solutions RSTV2143838
Encompass_Preferred
High Performance Enamel Spray Paint is the ultimate spray paint for industrial applications including metal, steel, masonry and wood. Advanced formula provides corrosion protection with easy, one-coat coverage and long-lasting color. High-output tip covers surfaces 50 percent faster and dries in only 20 minutes. Finished gloss is resistant to chipping, cracking and peeling and withstands heat up to 200 degrees Fahrenheit. Spray paint meets USDA requirements for incidental food contact and is VOC compliant nationwide.
Catalog No. 50-008-29084