Learn More
Selleck Chemical LLC Ruthenium Red E2993-50MG
SDP
Supplier:  Selleck Chemical LLC E299350MG
Encompass_Preferred
Ruthenium red (Ammoniated ruthenium oxychloride) is a polycationic dye which is a L-type calcium current (ICa) blocker It is widely used for electron microscopy (EM) of cells tissues and vegetative bacteria Ruthenium red strongly reacts with phospholipids and fatty acids and binds to acidic mucopolysaccharides
Catalog No. 50-000-40715