Learn More
Medchemexpress LLC RuvB E.coli His-SU 10ug
SDP
Supplier:  Medchemexpress LLC HYP7151310UG
Encompass_Preferred
RuvB Protein, E. coli (His-SUMO) is a recombinant Holliday junction ATP-dependent DNA helicase expressed in Escherichia coli with an N-terminal His-SUMO affinity tag. The full-length protein (≈336 amino acids, ~53.2 kDa) is provided in microgram-scale quantities for biochemical, structural, and protein interaction studies of DNA repair and recombination pathways.
  • Suitable for biochemical and structural studies.
  • Expressed in Escherichia coli for recombinant production.
  • N-terminal His-SUMO tag enables affinity purification and improved solubility.
  • Full-length sequence (~336 amino acids) preserves native activity.
  • Approximate molecular weight ~53.2 kDa for gel reference.
  • Provided in microgram-scale quantities for assay development and characterization.
Catalog No. 50-001-95902