Learn More
TARGETMOL CHEMICALS INC RUXOLITINIB 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T182950MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ruxolitinib (INCB018424) is a potent and selective JAK1/2 inhibitor (IC50 3.3/2.8 nM) and is relatively less selective for JAK3 (IC50 322 nM). purity: 99%
Catalog No. 50-257-7292