Learn More
Medchemexpress LLC Ruxolitinib | 941678-49-5 | MFCD12031592 | 99.7% | 306.37 g/mol | C17H18N6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY50856R10MG
Encompass_Preferred
Ruxolitinib analytical standard used as a reference material for research and analytical applications. It is a potent and selective JAK1/JAK2 inhibitor provided as a high-purity, small-quantity reference suitable for method development and quality control.
  • Analytical standard for method development and quality control.
  • High purity reference material (99.7%).
  • Molecular weight 306.37 g/mol; formula C17H18N6.
  • Intended for research and analytical applications.
  • Available in small, measured quantities suitable for analytical assays.
Catalog No. 50-002-98797