Learn More
Medchemexpress LLC Ruxolitinib impurity-1 | 952518-97-7 | MFCD20683197 | 97.0% | 185.19 g/mol | C9H7N5 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW059210
Ruxolitinib impurity-1 is an identified impurity of the Janus kinase inhibitor ruxolitinib supplied as an analytical reference standard for impurity profiling and safety analysis. The compound has CAS number 952518-97-7, molecular formula C9H7N5, and molecular weight 185.19 g/mol. It is provided in milligram to gram quantities for research and analytical use and is accompanied by a product datasheet and safety data sheet.
- Used as analytical reference standard for impurity profiling and safety analysis
- Cas number 952518-97-7
- Molecular formula C9H7N5
- Molecular weight 185.19 g/mol
- Available in milligram and gram quantities, e.g., 250 MG
- Accompanied by a product datasheet and safety data sheet
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.