Learn More
Selleck Chemical LLC Ruxolitinib (INCB018424) 100mg 941678-49-5
SDP
Supplier:  Selleck Chemical LLC S1378100MG
Encompass_Preferred
Ruxolitinib (INCB018424) is the first potent, selective, JAK1/2 inhibitor to enter the clinic with IC50 of 3.3 nM/2.8 nM in cell-free assays, >130-fold selectivity for JAK1/2 versus JAK3. Ruxolitinib kills tumor cells through toxic mitophagy. Ruxolitinib induces autophagy and enhances apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-847-3