Learn More
TARGETMOL CHEMICALS INC RUXOLITINIB PHOSPHATE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3043100MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ruxolitinib phosphate (INCB018424)(INCB018424 phosphate) is the first potent selective JAK1/2 inhibitor to enter the clinic with IC50 of 3.3 nM/2.8 nM >130-fold selectivity for JAK1/2 versus JAK3. purity: 99%
Catalog No. 50-258-0416