Learn More
Medchemexpress LLC HY-50858 1g , Ruxolitinib (phosphate) CAS:1092939-17-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY508581G
Encompass_Preferred
HY-50858 1g Ruxolitinib (phosphate) CAS: 1092939-17-7 Ruxolitinib phosphate (INCB018424 phosphate) is a potent JAK1/2 inhibitor with IC50s of 3.3 nM/2.8 nM, respectively, showing more than 130-fold selectivity over JAK3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9209