Learn More
Medchemexpress LLC Ruxolitinib (S enantiomer) | 941685-37-6 | 99.6% | C17H18N6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY50856A10MM/1ML
Encompass_Preferred
Ruxolitinib (S enantiomer), also known as S-Ruxolitinib, is the S-enantiomer of Ruxolitinib. It is an orally active and potent JAK inhibitor, targeting the JAK/STAT signaling pathway. This compound is supplied as a 10 mM solution in DMSO, ready for reconstitution.
  • Potent JAK inhibitor
  • Orally active compound
  • Suppresses IFNβ-induced gene expression in human small airway epithelial cells
  • Reduces haematopoietic cell expansion in myeloproliferative neoplasms in vivo
  • High solubility in DMSO (≥ 100 mg/mL)
  • Recommended storage for powder at -20°C for 3 years, and in solvent at -80°C for 2 years
Catalog No. 50-003-59306