Learn More
Medchemexpress LLC Ruxolitinib (Standard) | 941678-49-5 | MFCD12031592 | 99.74% | C17H18N6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY50856R
Encompass_Preferred
Ruxolitinib (Standard) is the analytical standard of Ruxolitinib, intended for research and analytical applications. It functions as a potent and selective JAK1/2 inhibitor, inducing autophagy and leading to the elimination of tumor cells through toxic mitophagy.
  • Potent and selective JAK1/2 inhibitor
  • Induces autophagy and kills tumor cells
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS
Catalog No. 50-003-57585