Learn More
MOLTOX Molecular Toxicology Inc Rappaport-Vassiliadis R10 Broth, 1000 mL/bottle, Store at Room Temperature
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 5140S119
Encompass_Preferred
Used for the isolation and identification of Salmonella and Shigella. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-848