Learn More
TARGETMOL CHEMICALS INC RWJ-51204 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T168115MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RWJ-51204 is A potent and selective adenosine A2A receptor antagonist. RWJ-51204 is a partial agonist of GABA(A) receptor (IC50 0.2-2 nM) which has neuroprotective effects and improves motor function and is a candidate compound for the treatment of Parkinson's disease. purity: 99%
Catalog No. 50-258-1148