Learn More
Cayman Chemical RWJ 52353hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1194210 MG
Encompass_Preferred
An α2D-AR agonist (Kis = 1.5, 254, 621, and 443 nM for α2D-, α2A-, α2B-, and α1-ARs, respectively); has antinociceptive properties in the mouse and rat abdominal irritation tests (ED50s = 11.6 and 15.1 mg/kg, respectively)
Catalog No. 50-295-4620