Learn More
Medchemexpress LLC RX 801077 (hydrochloride) | 89196-95-2 | MFCD00672666 | 99.8% | 222.6708 | C11H11ClN2O | 10 MG

Supplier: Medchemexpress LLC HY10090410MG
RX 801077 hydrochloride is a selective imidazoline I2 receptor (I2R) agonist provided for research use as a solid or as a 10 mM solution in DMSO. It is produced with high purity for pharmacological studies, including investigations of neuroprotection and anti-inflammatory mechanisms.
- Selective imidazoline I2 receptor agonist with reported Ki ≈ 70 nM.
- High purity, 99.8% as supplied.
- Available as solid and as a 10 mM solution in DMSO.
- Suitable for pharmacology, neuroprotection, and inflammation research.
- Store at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
- Soluble in DMSO for stock solution preparation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.