Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ryl-552 | 1801444-56-3 | 98.6% | 1 ML
SDP

Supplier:  Medchemexpress LLC HY12033810MM 1ML

Encompass_Preferred

RYL-552 is a research-grade small molecule inhibitor of the mitochondrial electron transport chain that targets Plasmodium falciparum NADH dehydrogenase 2 (PfNDH2). Supplied as solid material and as a 10 mM solution in DMSO for in vitro research use.

  • PfNDH2 inhibitor active against P. falciparum
  • High purity: 98.6% (as supplied)
  • Available as solid in multiple milligram quantities and as 10 mM in DMSO, 1 mL
  • Soluble in DMSO: 50 mg/mL (≈116.99 mM)
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years)
  • In solvent storage: -80°C (up to 6 months) or -20°C (up to 1 month)

Catalog No. 50-002-33445


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.