Learn More
Electron Microscopy Sciences Double Edge Razor Blades Non-Wax Type
SDP
Supplier:  Electron Microscopy Sciences 7200205
Encompass_Preferred
Made from carbon steel. Feather blades are have super sharp edges. Easily breaks in half for use with any tissue slicing machine, or break smaller portions for use as a microscalpel with a Blade Breaker & Holder. With a clean non-wax surface.
Catalog No. 50-192-7974