Learn More
Cayman Chemical S- 1 1-BI-2-naphthol 5g
SDP
Supplier:  Cayman Chemical 700525 G
Encompass_Preferred
A chiral auxiliary for asymmetric ketone reduction, in particular 15-keto PG intermediates to 15(S)-alcohols
Catalog No. 50-289-6880