Learn More
Medchemexpress LLC (S)-(+)-1,2-Propanediol | 4254-15-3 | 99.8% | C₃H₈O₂ | 100 G
SDP
Supplier:  Medchemexpress LLC HY79334100G
Encompass_Preferred
(S)-(+)-1,2-Propanediol is an endogenous metabolite that can be used as an organic solvent and diluent in drug preparations. It also serves as a chiral synthetic intermediate and a substrate for enzyme research.
  • Used as an organic solvent and diluent
  • Serves as a chiral synthetic intermediate
  • Acts as a substrate for enzyme research
  • Endogenous metabolite
Catalog No. 50-004-32685