Learn More
Medchemexpress LLC (S)-1-(3-(benzyloxy)isoxazol-5-yl)-N,N-dimethylprop-2-en-1-amine | 951698-15-0 | 99.1% | C15H18N2O2 | 10MG

Supplier: Medchemexpress LLC HY13588810MG
TP748 is an isoxazole building block used as a key intermediate in the chemical synthesis of fully synthetic tetracycline derivatives. Supplied for research use, it is provided as a high-purity solid suitable for synthetic and medicinal chemistry workflows.
- High purity (99.1%).
- White to off-white solid.
- Molecular formula C15H18N2O2; molar mass 258.32 g/mol.
- Soluble in DMSO (100 mg/mL).
- Powder stable at -20°C for long-term storage.
- Designed for use as a synthetic intermediate in tetracycline chemistry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.