Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-1-benzylpyrrolidin-3-ol | 101385-90-4 | MFCD00075485 | 177.25 | C11H15NO | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW016644

Encompass_Preferred

(S)-1-benzylpyrrolidin-3-ol is a chiral pyrrolidine intermediate used in pharmaceutical synthesis, notably for preparing impurities of the antihypertensive barnidipine hydrochloride. The compound has molecular formula C11H15NO, molecular weight 177.25, and CAS 101385-90-4. Manufacturer datasheet reports solubility and storage conditions and lists small-scale package sizes for laboratory use.

  • Chiral pyrrolidine building block for drug synthesis.
  • Used to synthesize barnidipine hydrochloride impurities.
  • Molecular formula C11H15NO; molecular weight 177.25.
  • Solubility: DMSO 200 mg/mL; in vivo formulation examples provided.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available package sizes: 5 g, 10 g, 25 g.

Catalog No. 50-003-03822


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.