Learn More
Cayman Chemical S-1 MethndamIde 10mg
SDP
Supplier:  Cayman Chemical 9007210 MG
Encompass_Preferred
A CB1 receptor ligand with less potency than R-1 methanandamide; inhibits electrically evoked contractions in isolated murine vas deferens (IC50 = 230 nM); binding affinity for CB1 receptors is less than that of AEA (Ki = 175 nM for the displacement of radiolabeled CP 55,940)
Catalog No. 50-289-5078