Learn More
Medchemexpress LLC S-1-Propenyl-L-cysteine | 52438-09-2 | 99.9% | 161.22 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111827
Encompass_Preferred
S-1-Propenyl-L-cysteine is a stereoisomer of S-allyl-L-cysteine, extracted from garlic. It has immunomodulatory effects and reduces blood pressure in a hypertensive animal model. It exhibits antioxidative efficacy through a NO-dependent BACH1 signaling pathway and is orally active.
  • Immunomodulatory effects
  • Reduces blood pressure in a hypertensive animal model
  • Exhibits antioxidative efficacy through a NO-dependent BACH1 signaling pathway
  • It is orally active
  • Upregulates HMOX1
  • Inhibits TLR4 signaling
  • Reduces systolic blood pressure (SBP)
  • Alters histidine metabolism via the central histamine H3 receptor
Catalog No. 50-004-38732