Learn More
Medchemexpress LLC S-14506 hydrochloride | 286369-38-8 | 99.9% | 443.94 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11002450MG
Encompass_Preferred
S-14506 hydrochloride is a potent 5-HT1A agonist and a 5-HT2A/2C antagonist. It also exhibits dopamine antagonist properties by blocking dopamine D2 receptors, inhibiting the in vivo binding of [3H]raclopride in the striatum and olfactory bulbs. This compound has potential for research as an anxiolytic agent.
  • Potent 5-HT1A agonist
  • 5-HT2A/2C antagonist
  • Exhibits dopamine antagonist properties by blocking dopamine D2 receptors
  • Inhibits in vivo binding of [3H]raclopride
  • Potential for anxiolytic agent research
  • High affinity for G protein-uncoupled state of 5-HT receptor in HeLa cells
  • Dose-dependently antagonizes apomorphine-induced stereotyped climbing and sniffing in mice
  • Active in the forced swimming test in rats, inducing immobility
Catalog No. 50-003-44992