Learn More
Medchemexpress LLC (S)-2-(1,4-dibenzylpiperazin-2-yl)acetonitrile | 2225787-92-6 | 99.9% | 305.43 | C20H23N3 | 100 MG

Supplier: Medchemexpress LLC HYW049455100MG
(S)-2-(1,4-dibenzylpiperazin-2-yl)acetonitrile is a chiral drug intermediate used in the synthesis of active pharmaceutical compounds and for laboratory research applications.
- High purity (99.9% by LCMS).
- Molecular formula C20H23N3, molecular weight 305.43.
- Identified uses: laboratory chemicals and manufacture of substances.
- Appearance oil; reported boiling point ≈462.6 ± 30.0 °C (760 mmHg).
- Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.