Learn More
Medchemexpress LLC (S)-2-(2-(1H-Indol-3-yl)acetamido)-3-phenylpropanoic acid | 57105-50-7 | 97.2% | 322.36 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0112805G
Encompass_Preferred
(S)-2-(2-(1H-Indol-3-yl)acetamido)-3-phenylpropanoic acid is a phenylalanine derivative. Amino acids and amino acid derivatives, such as this product, have been commercially used as ergogenic supplements. They are known to influence the secretion of anabolic hormones, supply fuel during exercise, enhance mental performance during stress-related tasks, and prevent exercise-induced muscle damage. This product is for research use only and not sold to patients.
  • Phenylalanine derivative
  • Supports ergogenic supplementation
  • Influences anabolic hormone secretion
  • Provides fuel during exercise
  • Enhances mental performance
  • Helps prevent exercise-induced muscle damage
  • Appears as a white to off-white solid
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years
  • Can be stored in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-003-66544