Learn More
Medchemexpress LLC (S)-2-(2-((3'-(1-amino-2-hydroxyethyl)-[1,1'-biphenyl]-3-yl)methoxy)phenyl)acetic acid | 1646682-14-5 | 99.2% | 377.43 g/mol | C23H23NO4 | 5 MG

Supplier: Medchemexpress LLC HY1285705MG
FD-IN-1 is a small-molecule research compound that selectively inhibits complement Factor D (FD). It demonstrates potent biochemical activity and functional inhibition in whole-blood assays, and is supplied as a solid or DMSO stock for use in in vitro and in vivo preclinical studies.
- Selective factor D inhibition with IC50 of 12 nM.
- Functional inhibition in MAC deposition whole-blood assay (IC50 0.26 μM).
- High purity suitable for research (≈99.2% reported).
- Available as solid samples and as a DMSO stock solution for flexible use.
- Soluble in DMSO and compatible with common in vivo formulation vehicles.
- Recommended storage conditions for powder and stock solutions to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.