Learn More
Strem, An Ascensus Company CAS# 1365531-82-3. 100mg. (S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, min. 97% (S)-Tol-Garphos™. MFCD19443623
SDP
Supplier:  Strem, An Ascensus Company 151658100MG
Encompass
CAS# 1365531-82-3. 100mg. (S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, min. 97% (S)-Tol-Garphos™. MFCD19443623. Molecular Weight: 698.77. Molecular Formula: C44H44O4P2. Color/form: white xtl. Strem# 15-1658. http://www.strem.com/catalog/v/15-1658/
Catalog No. 50-369-8