Learn More
Strem, An Ascensus Company CAS# 139139-93-8. 250mg. (S)-(-)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl (S)-(-)-H8-BINAP. MFCD01630795
SDP
Supplier:  Strem, An Ascensus Company 152973250MG
Encompass
CAS# 139139-93-8. 250mg. (S)-(-)-2,2'-Bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl (S)-(-)-H8-BINAP. MFCD01630795. Molecular Weight: 630.74. Molecular Formula: C44H40P2. Color/form: off-white pwdr. Strem# 15-2973. http://www.strem.com/catalog/v/15-2973/
Catalog No. 50-901-10251