Learn More
Medchemexpress LLC Glycine, l-γ-glutamyl-S-(2,4-dinitrophenyl)-l-cysteinyl- | 26289-39-4 | MFCD01863384 | 99.9% | C16H19N5O10S | 10MG
Supplier: Medchemexpress LLC HY13733010MG
S-(2,4-Dinitrophenyl)glutathione is a glutathione conjugate in which the thiol of glutathione is substituted with a 2,4-dinitrophenyl group. Supplied as a high-purity powder, it is used as a substrate in glutathione-S-transferase (GST) activity assays, enzyme kinetics studies, and related biochemical research.
- High-purity powder (99.9%) suitable for analytical applications.
- Reliable substrate for glutathione-S-transferase activity assays.
- Well-characterized molecular composition for reproducible results.
- Stable when stored sealed and protected from moisture.
- Available in small quantities for research-scale experiments.