Learn More
Medchemexpress LLC S-2-(4-isothiocyanatobenzyl)-1,4,7,10-tetraazacyclododecane tetraacetic acid | 1020407-41-3 | 99.8% | 551.61 g/mol | C24H33N5O8S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15603810MG
Encompass_Preferred
(S)-p-SCN-Bn-DOTA is a bifunctional DOTA-derived chelator used in radiopharmaceutical research. It incorporates an isothiocyanatobenzyl reactive group for covalent attachment to biomolecules and a macrocyclic DOTA core for stable radiometal coordination.
  • Enables stable chelation of radiometals for imaging and therapy.
  • Provides an isothiocyanate reactive handle for covalent conjugation to proteins and peptides.
  • Suitable for labeling studies in small research quantities.
  • High chemical purity supports reproducible conjugation and radiolabeling.
  • Solid form with recommended cold storage to maintain stability.
Catalog No. 50-002-88129