Learn More
Medchemexpress LLC (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(o-tolyl)propanoic acid | 211637-75-1 | 99.9% | 401.45 | 5 G

Supplier: Medchemexpress LLC HYW0109845G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(o-tolyl)propanoic acid is a phenylalanine derivative. Amino acids and amino acid derivatives influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage powder: -20°C 3 years, 4°C 2 years
- Storage in solvent: -80°C 6 months, -20°C 1 month
- Influence the secretion of anabolic hormones
- Supply of fuel during exercise
- Mental performance during stress-related tasks
- Prevent exercise-induced muscle damage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.