Learn More
Medchemexpress LLC (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-oxo-4-(tritylamino)butanoic acid | 941296-80-6 | MFCD22200773 | ≥97.0% | 610.70 g/mol | C39H34N2O5 | 1 G

Supplier: Medchemexpress LLC HYW1303971G
An Fmoc-protected N-methyl asparagine derivative used as a research reagent and building block for solid-phase peptide synthesis (SPPS) and related organic synthesis. It contains an Fmoc N-terminal protecting group and a trityl-protected side chain to enable selective coupling and deprotection during peptide assembly.
- Suitable for solid-phase peptide synthesis and peptide modification.
- Fmoc-protected N-methyl asparagine with trityl side-chain protection.
- Molecular weight 610.70 g/mol; formula C39H34N2O5.
- Reported purity typically ≥97.0% by HPLC from commercial sources.
- Storage: powder -20°C (long term) or 4°C; in solvent -80°C or -20°C.
- Supplied in small research pack sizes for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.