Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((1r,4S)-4-methylcyclohexyl)acetic acid | 50 g | CAS 1312298-38-6 | InChIKey HKFDJGRKUVMKLM-WCJKSRRJSA-N

Supplier:  STA PHARMACEUTICAL US LLC STAAA1221050GM

Encompass

(S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((1r,4S)-4-methylcyclohexyl)acetic acid is a Amino Acid reagent (Subcategory: Alkane AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1312298-38-6
- MDL: No data
- InChIKey: HKFDJGRKUVMKLM-WCJKSRRJSA-N
- Molecular Weight: 393.483
- Molecular Formula: C24H27NO4
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((1r,4S)-4-methylcyclohexyl)acetic acid
- SMILES: C[C@H]1CC[C@@H](CC1)[C@@H](C(O)=O)NC(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O

Catalog No. 50-235-3890


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.