Learn More
Medchemexpress LLC (S)-2-allylpyrrolidine-2-carboxylic acid hydrochloride | 129704-91-2 | 98.0% | C8H14CINO2 | 250 MG

Supplier: Medchemexpress LLC 250MGHY75869
(S)-2-allylpyrrolidine-2-carboxylic acid hydrochloride is a chemical substance primarily utilized as a laboratory chemical and a crucial drug intermediate for the synthesis of various active compounds. This product is presented as a solid, with a melting point ranging from 186-190 °C, and boasts a high purity of 98.0%.
- Serves as a drug intermediate
- Suitable for laboratory chemical applications
- Solid form for ease of handling
- High purity of 98.0%
- Stable as powder at -20°C for up to 3 years
- Stable as powder at 4°C for up to 2 years
- Stable in solvent at -80°C for up to 6 months
- Stable in solvent at -20°C for up to 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.