Learn More
Medchemexpress LLC (S)-2-amino-2-(thiophen-2-yl)acetic acid | 43189-45-3 | MFCD00078081 | 98.0% | 157.19 | C6H7NO2S | 100 MG

Supplier: Medchemexpress LLC HYW017256100MG
(S)-2-Amino-2-(thiophen-2-yl)acetic acid is a chiral amino-acid derivative of glycine supplied as a white to off-white solid for research use. It has molecular formula C6H7NO2S, molecular weight 157.19, and CAS number 43189-45-3. The compound is used in biochemical and pharmacological studies where a thiophene-substituted glycine analogue is required; refer to the supplier datasheet and SDS for handling and safety information.
- White to off-white solid physical form.
- Molecular formula C6H7NO2S; molecular weight 157.19.
- Suitable for biochemical and pharmacological research applications.
- Storage: 4°C; in solvent: -80°C for 6 months, -20°C for 1 month; protect from light.
- Typical purity reported as 98.0% by distributors.
- For research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.