Learn More
Medchemexpress LLC (S)-2-amino-3-(3-nitrophenyl)propanoic acid | 19883-74-0 | MFCD01860661 | 100.0% | 210.19 g/mol | C9H10N2O4 | 25 G

Supplier: Medchemexpress LLC 25GHYW050494
(S)-2-Amino-3-(3-nitrophenyl)propanoic acid is the S-enantiomer of a nitro-substituted phenylalanine derivative supplied as a white to off-white solid for research use. It is used as a research reagent in amino-acid chemistry and biochemical studies and is available in laboratory pack sizes for experimental and analytical applications.
- S-enantiomer nitro-substituted phenylalanine derivative
- White to off-white solid powder
- Molecular weight 210.19 g/mol; formula C9H10N2O4
- High purity (99.96%)
- Available in laboratory pack sizes, including 25 g
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.