Learn More
Medchemexpress LLC (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate | 18835-59-1 | 469.01 | 1 ML

Supplier: Medchemexpress LLC HYW00843710MM/1ML
This product is (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate, an endogenous metabolite. It is supplied as a solid, with an appearance ranging from white to yellow, and is intended for research use only.
- Endogenous metabolite
- Purity: 99.0%
- Molecular weight: 469.01
- Chemical formula: C9H13I2NO5
- Appearance: Solid, white to yellow
- Shipping: Room temperature
- Storage: -20°C, sealed, away from moisture
- Solubility in DMSO: 100 mg/mL
- Classified as natural product and phenol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.