Learn More
Medchemexpress LLC (S)-2-amino-3-(benzo[b]thiophen-3-yl)propanoic acid | 72120-71-9 | MFCD00079684 | 221.28 g·mol⁻¹ | C11H11NO2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW022796100MG
Encompass_Preferred
(S)-2-Amino-3-(benzo[b]thiophen-3-yl)propanoic acid is the S-enantiomer of a benzo[b]thiophene-substituted alanine. It is a chiral, heteroaryl-containing amino acid used as a building block in medicinal chemistry, peptide synthesis, and biochemical research. Supplied as a solid for research use only.
  • Chiral S-enantiomer for stereospecific studies.
  • Benzo[b]thiophene side chain provides heteroaryl functionality.
  • Suitable as a building block in peptide synthesis and small-molecule design.
  • Solid form; molecular formula C11H11NO2S; molecular weight 221.28 g·mol⁻¹.
  • Intended for research use only.
Catalog No. 50-002-99611